Skip to Content
Merck
All Photos(1)

Key Documents

11483188001

Roche

First Strand cDNA Synthesis Kit for RT-PCR (AMV)

sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included

Synonym(s):

RT-PCR, cDNA sysnthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106313

usage

sufficient for 30 reactions (including 5 control reactions)

Quality Level

specific activity

>50 units/μg protein

feature

dNTPs included
hotstart: no

packaging

kit of 1 (10 components)

manufacturer/tradename

Roche

parameter

42 °C optimum reaction temp.

technique(s)

RT-PCR: suitable

input

purified RNA

storage temp.

−20°C

General description

The First Strand cDNA Synthesis Kit is used for the synthesis of the first strand cDNA as the starting reaction for two-step RT PCR.The kit includes Reverse Transcriptase AMV for first strand synthesis, two different primers, our PCR Nucleotide Mix, and Control Neo pa RNA. PCR products that are generated by RT-PCR can be cloned using standard procedures.
The amplification of RNA requires the conversion of the RNA substrate into DNA. This is achieved through the use of a reverse transcriptase such as AMV RT (avian myeloblastis virus reverse transcriptase) or M-MuLV RT (moloney murine leukemia virus reverse transcriptase). The resulting cDNA can be used as a template for a standard PCR.
AMV RT synthesizes the new cDNA strand at site(s) determined by the type of the primer used:
  • at the 3′-end of the poly(A) mRNA when Oligo-p(dT)15is used as a primer,
  • at nonspecific points along the mRNA template when using the random primer p(dN)6, or
  • at a site determined by a sequence-specific primer.
The use of AMV RT for first-strand cDNA synthesis has certain advantages. The higher thermostability of this enzyme compared to M-MuLV RT allows the reaction to be performed at +42°C, giving higher specificity and better resolution of secondary structures compared to a reaction performed at +37°C.

Specificity

Heat inactivation: 5 min, 95 °C
Reverse Transcriptase AMV

Application

The First Strand cDNA Synthesis Kit for RT-PCR (AMV) is suitable for:
  • Detection of the presence or absence of RNA viruses or other RNA-containing microorganisms (in combination with PCR)
  • Quantification of mRNA for monitoring differential expression of a specific mRNA
  • First step in the "differential display of mRNA"
  • Generation of cDNA libraries with large and full-length inserts
  • Reverse transcription-quantitative polymerase chain reaction (RT-qPCR) for reverse transcription of RNA into complementary DNA.

Features and Benefits

Robustness:
  • Transcribes total RNA, mRNA and viral RNA along with difficult-to-transcribe secondary RNA structures
  • Obtain cDNA transcripts up to 12 kb
  • Higher thermostability (up to 50°C) and specificity than M-MuLV Reverse Transcriptase
  • Purification of the resulting cDNA before PCR reaction is not necessary
Flexibility:
It can be used with either sequence-specific primers, poly(dT)15 primers, or random primers, p(dN)6

Packaging

1 kit containing 10 components.

Quality

In the standard cDNA synthesis assay, at least 300 ng of cDNA is synthesized (i.e., 30% yield) when 1.0 μg Neo pa RNA template is incubated with 20 μCi [α-32P]-dCTP (specific activity of 3000 Ci/mmol) for 60 minutes at +42°C.

Unit Definition

Volume Activity: 20 to 25 U/μl

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Reverse Transcriptase AMV

  • PCR Nucleotide Mix, pH 8.5 10 mM each

  • Reaction Buffer

  • MgCl2 Stock Solution 25 mM

  • Gelatin 0.05% (w/v)

  • Oligo-p(dT)15 , 0.02 A260 units/μl 0.8 μg/μl

  • Random Primer p(dN)6 , 0.04 A260 units/μl 1.6 μg/μl

  • RNase Inhibitor 50 U/μl

  • Control Neo pa RNA (1.0 kb in length with additional 19-base 3′-poly(A) tail) 0.2 μg/μl

  • Water, PCR Grade

See All (10)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Han et al.
Molecular medicine reports, 18(1), 1058-1066 (2018-05-31)
Atorvastatin is a member of the statin class of drugs, which competitively inhibit the activity of 5‑hydroxy‑3‑methylglutaryl‑coenzyme A reductase. The aim of the present study was to assess whether atorvastatin may protect BV‑2 microglia and hippocampal neurons against oxygen‑glucose deprivation
Screening of Selected Soil and Endophytic Fungi for Lovostatin
Biosynthetic Genes lovE and lovF
Bhargavi SD
Microbial Biotechnology (2015)
Relationship of Podoplanin and Glutathione S-transferases T1 Expression with Laryngeal Cancer
Deniz K
International journal of molecular and cellular medicine (2012)
Roles of Signal Transducer Pathways in Investigation of Biopsies from Patients with Bladder Tumors
Aysegul Bayrak
Asian Pacific Journal of Cancer Prevention (2017)
Bita Nickkholgh et al.
Fertility and sterility, 102(2), 558-565 (2014-05-28)
To evaluate the degree of enrichment of spermatogonial stem cells (SSCs) from human testicular cell cultures by ITGA6+, HLA-/ITGA6+, GPR125+, and HLA-/GPR125+ magnetic-assisted cell sorting (MACS). Experimental basic science study. Reproductive biology laboratory. Multiple samples of cryopreserved human testicular cells

Articles

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Protocols

Related Content

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

RT-qPCR detects specific targets with applications in gene expression and pathogen detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service