Skip to Content
Merck
All Photos(1)

Key Documents

EX0190

Supelco

Diethyl ether

≥99.0% (GC), anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Diethyl ether, Diethyl Ether, Ether, Diethyl Ether, Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12352112
EC Index Number:
200-467-2
Assay:
≥99.0% (GC)
bp:
34.6 °C (lit.)
vapor pressure:
587 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Ethyl Ether, anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

vapor density

2.6 (vs air)

Quality Level

vapor pressure

587 hPa ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

320 °F

shelf life

24 mo. (from mfg date)

expl. lim.

36.5 %

impurities

≤0.0002 meq/g Titrable acid
≤0.001% Carbonyl (as HCHO)
≤0.03% Water (H2O)
≤1.0 ppm Peroxide (as H2O2)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

solubility

69 g/L

density

0.706 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

2-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl ether (CH3CH2)2O is also referred to as sulfuric ether, diethyl ether, or just ether is used as a laboratory solvent. Ether is a desirable solvent for oils, fats, waxes, perfumes, dyes, resins, hydrocarbons, and gums. It is also used as fuel and to synthesize other chemical compounds.

Application

Ethyl ether can be used as:
  • A solvent in organic reactions such as Grignard and Wurtz reactions.
  • A solvent along with lithium perchlorate (LPDE) in the reductive monoalkylation of amines and its derivative with aldehyde.
  • A solvent for the extraction and purification of (L+) lactic acid from fermented broth and synthesize PLA (polylactic acid).

Analysis Note

Expiration date: Exp 24 months from mfg date
Alcohol(C2H5OH) (limit about 0.05%): To pass test
Assay (GC): 99.0% min
Carbonyl (as HCHO): 0.001% max
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Peroxide (as H2O2): 1.0ppm max
Titrable acid: 0.0002meq/g max
Water (H2O): 0.03% max

Stabilized with ≥1ppm BHT

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service