Skip to Content
Merck
All Photos(1)

Key Documents

EX0182

Supelco

Diethyl ether

≥99.9% (GC), suitable for LC/MS, gas chromatography (GC), HPLC, OmniSolv®

Synonym(s):

Diethyl ether, Diethyl Ether, Ether, Diethyl Ether, Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12352112
EC Index Number:
200-467-2
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
34.6 °C (lit.)
vapor pressure:
587 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Ethyl Ether, OmniSolv®

vapor density

2.6 (vs air)

Quality Level

vapor pressure

587 hPa ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

320 °F

shelf life

12 mo. (from manufacture date)

expl. lim.

36.5 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.030% Water (H2O)
≤1 ppm Peroxide (as H2O2) (at time of packaging)

evapn. residue

≤1.0 ppm

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

solubility

69 g/L

density

0.706 g/mL at 25 °C (lit.)

UV cutoff

≤212 nm

UV absorption

λ: 212 nm Amax: ≤1.000
λ: 250 nm Amax: ≤0.080
λ: 275 nm Amax: ≤0.010
λ: 300 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

shipped in

ambient

storage temp.

2-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

EX0182-1 is packaged in poly coated glass bottles
Unstabilized/Expiration Dated
OmniSolv®

Analysis Note

Expiration dateExp 12 months from mfg dateAppearanceClear, free from particulatesAssay (GC)99.9% minCapillary ECD responsive substances (as PCNB)10ng/L maxColor (APHA)10 maxEvaporation residue1.0ppm maxFiltered for particulate matterTo pass testFormClear liquidPeroxide (as H2O2) (at time of packaging)1ppm maxRefractive index (at 20°C)1.3471-1.3571UV Abs. at 212 nm1.000AU maxUV Abs. at 250 nm0.080AU maxUV Abs. at 275 nm0.010AU maxUV Abs. at 300 nm0.005AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off212nm maxWater (H2O)0.030% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service