1.02439
Eosin Y-solution 0.5% alcoholic
for microscopy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116124
NACRES:
NA.41
Pricing and availability is not currently available.
Quality Level
form
liquid
IVD
for in vitro diagnostic use
technique(s)
microbe id | staining: suitable
transition temp
flash point 19.1 °C
density
0.8889 g/cm3 at 20 °C
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
The Eosin Y-solution 0.5%, alcoholic - for microscopy, is a ready-to-use solution that can be used for routine staining in the hematoxylin-eosin staining process (H&E) of sample material of human origin. The H&E staining process is the most frequently used staining method for histological material. The staining mechanism behind the H&E staining is a physico-chemical process. In the first step, the positively charged nuclear dye (Mayer′s hematoxylin , Product number 1.09249, or Hematoxylin solution modified acc. to Gil III, Product number 1.05154) binds to the negatively charged phosphate groups of the nucleic acid of the cell nucleus. The nuclei will be dyed dark blue to dark violet. The second step is the counterstaining with negatively charged anionic Eosin Y, a xanthene dye. Eosin binds to the positively charged plasma proteins. Cytoplasma and intercellular substances are stained pink to red, while erythrocytes will appear with yellow to orange color.
The 500 ml bottle is sufficient for approx. 750 specimens. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
The 500 ml bottle is sufficient for approx. 750 specimens. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
66.4 °F
Flash Point(C)
19.1 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service