Skip to Content
Merck
All Photos(1)

Key Documents

8.02921

Sigma-Aldrich

1,6-Dibromohexane

for synthesis

Synonym(s):

1,6-Dibromohexane, Hexamethylene bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)6Br
CAS Number:
Molecular Weight:
243.97
MDL number:
UNSPSC Code:
12352101
EC Index Number:
211-067-2
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

potency

1800 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

mp

-85 °C

transition temp

flash point >110 °C

density

1.61 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCCCCCCBr

InChI

1S/C6H12Br2/c7-5-3-1-2-4-6-8/h1-6H2

InChI key

SGRHVVLXEBNBDV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.604 - 1.606
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service