Skip to Content
Merck
All Photos(3)

Documents

570958

Sigma-Aldrich

Tri-tert-butylphosphine

98%

Synonym(s):

P(t-Bu)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]3P
CAS Number:
Molecular Weight:
202.32
Beilstein:
1738613
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

bp

102-103 °C/13 mmHg (lit.)

mp

30-35 °C (lit.)

density

0.834 g/mL at 20 °C (lit.)

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P/c1-10(2,3)13(11(4,5)6)12(7,8)9/h1-9H3

InChI key

BWHDROKFUHTORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Preferred ligand in the palladium-catalyzed coupling of arylboronic acids with phenyliodonium ylides of hydroxyquinones. Also used with Pd2(dba)3 for the α-arylation of trimethylsilyl enol ethers with aryl halides.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuo Iwama et al.
Organic letters, 8(25), 5725-5728 (2006-12-01)
Inter- and intramolecular arylations of trimethylsilyl enol ethers with aryl halides are accomplished regiospecifically in the presence of a palladium catalyst and tributyltin fluoride in refluxing benzene or toluene. The optimal catalyst system called for the use of Pd2(dba)3 and
Synlett, 2597-2597 (2006)
Maria V Zakharova et al.
ACS applied materials & interfaces, 10(15), 13199-13210 (2018-03-10)
Conventional amines and phosphines, such as diethylenetriamine, diphenylpropylphosphine, triethylamine, and tetramethylpiperidine, were grafted or impregnated on the surface of metalated SBA-15 materials, such as Ti-, Al-, and Zr-SBA-15, to generate air-stable solid-supported Lewis acid-base pairs. The Lewis acidity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service