Skip to Content
Merck
All Photos(3)

Key Documents

128007

Sigma-Aldrich

1,5-Dibromopentane

97%

Synonym(s):

Pentamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)5Br
CAS Number:
Molecular Weight:
229.94
Beilstein:
1209245
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

110 °C/15 mmHg (lit.)

mp

−34 °C (lit.)

density

1.688 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCCCCBr

InChI

1S/C5H10Br2/c6-4-2-1-3-5-7/h1-5H2

InChI key

IBODDUNKEPPBKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,5-Dibromopentane is a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast.

Application

1,5-Dibromopentane can be used to prepare:
  • Derivatives of 5-pyrazolones.
  • O-alkylated phloroglucinol derivatives, which forms the core of poly(alkyl aryl ether) dendrimers.
  • Intermediate for dezocine viz, (R)-(+)-1-(5-bromopentyl)-1-methyl-7-methoxy-2-tetralone using 1-methyl-7-methoxy-2-tetralone.

1,5-Dibromopentane was used in the preparartion of 1,5-di-Grignard reagent. It was also used in the synthesis of novel spirocyclic pyrrolidones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc. Perkin Trans. II, 2217-2217 (1992)
Enantioselective phase-transfer catalyzed alkylation of 1-methyl-7-methoxy-2-tetralone: an effective route to dezocine
Li R, et al.
Beilstein Journal of Organic Chemistry, 14(1), 1421-1427 (2018)
Ashraf Khan et al.
Journal of chromatographic science, 59(2), 112-119 (2020-12-09)
A new, simple and stability-indicating gas chromatography-flame ionization detection (GC-FID) method was developed and validated for the quantitative determination of busulfan and its organic impurities (OI) in drug substance without derivatization. The chromatographic attributes were achieved on a fused silica
Zhengxin Cai et al.
Nuclear medicine and biology, 43(1), 108-115 (2015-11-26)
Imaging fatty acid uptake and utilization has broad impact in investigating myocardial diseases, hepatic functions, tumor progression, and the metabolic state of adipose tissue. The SPECT tracer (123)I-15-(p-iodophenyl)-3-(R,S)-methylpentadecanoic acid (BMIPP) is a clinically used nuclear medicine tracer to image myocardial
Journal of Organometallic Chemistry, 466, 5-5 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service