Skip to Content
Merck
All Photos(2)

Key Documents

1.15440

Supelco

Isooctane

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

2,2,4-Trimethylpentane, 2,2,4-Trimethylpentane, Isobutyltrimethylmethane, iso-Octane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12191502
EC Index Number:
208-759-1
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)

vapor density

3.9 (vs air)

Quality Level

vapor pressure

41 mmHg ( 21 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

745 °F

potency

>2500 mg/kg LD50, oral (Rat)

expl. lim.

6 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

solubility

0.56 mg/L

density

0.692 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detector) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.

Application

Ideal for GC applications including sensitive detection in residue and environmental analysis.
The product is reported for used as a solvent in analyses of:
  • commercial technical mixtures (aroclors) and in environmental samples for polychlorinated dibenzo-p-dioxins (PCDD), polychlorinated dibenzofurans (PCDF) and polychlorinated biphenyls (PCB) by high resolution gas chromatography coupled to electron capture detector (HRGC-ECD)
  • sewage and soil samples for PCB congeners using HRGC-ECD or HRGC in conjunction with high resolution mass spectrometry (HRMS)

Specifications

Saturation Concentration in Air: 239 g/m3 (20 °C)

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of non-ortho polychlorinated biphenyl congeners on pre-packed carbon tubes. Application to analysis in sewage sludge and soil samples
Molina L, et al.
Chemosphere, 40(9-11), 921-927 (2000)
Fractionation of polychlorinated dibenzo-p-dioxins, polychlorinated dibenzofurans and planar polychlorinated biphenyls by high performance liquid chromatography on a pyrenyl-silica column
Martinez-Cored M, et al.
Fresenius Journal of Analytical Chemistry, 364(6), 576-583 (1999)

Articles

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service