Skip to Content
Merck
All Photos(2)

Key Documents

1.06011

Supelco

Methanol

≥99.8% (GC), suitable for gas chromatography (GC), SupraSolv®

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, for gas chromatography ECD and FID SupraSolv®

vapor density

1.11 (vs air)

Quality Level

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv ECD and FID are specially developed for gas chromatography in combination with the ECD (Electron Capture Detector) and FID (Flame Ionization Detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline, and a minimal signal-to-noise ratio, leading to reliable and reproducible analysis results.

Application

Methanol is used as a:
  • Solvent for preparing samples for GC/MS analysis of pyrolysis oil.[1]
  • Component of the solvent mixture used to extract oil samples.[2]

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flash pyrolysis of heavy metal contaminated hardwoods from phytoremediation: Characterisation of biomass, pyrolysis oil and char/ash fraction
Stals M, et al.
Journal of Analytical and Applied Pyrolysis, 89(1), 22-29 (2010)
Effects of repeated heating on fatty acid composition of plant-based cooking oils
Szabo Z, et al.
Food, 192, 11(2)-11(2) (2022)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service