Skip to Content
Merck
All Photos(2)

Key Documents

1.05587

Supelco

Sodium hydroxide solution

32%, for analysis EMPLURA®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sodium hydroxide solution about 32%, EMPLURA®

vapor pressure

3 mmHg ( 37 °C)
8 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

form

liquid

concentration

32.0-33.0% NaOH

impurities

≤0.0005% Heavy metals (as Pb)
≤0.0005% Phosphate (PO4)
≤0.001% Al (Aluminium)
≤0.001% Fe (Iron)
≤0.003% Sulfate (SO4)
≤0.004% Chloride (Cl)
≤0.005% Silicate (SiO2)

pH

14 (20 °C in H2O)

mp

9 °C

density

1.35 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Decomposition of sodium peroxybenzoate in sodium hydroxide solution.: This study investigates the decomposition process of sodium peroxybenzoate in sodium hydroxide solution. The research provides detailed insights into the chemical reactions and kinetics involved, which are essential for understanding the stability and reactivity of peroxy compounds in alkaline solutions (Tetrahedron, 1970).

Analysis Note

Assay (acidimetric,NaOH): 32.0 - 33.0 %
Carbonate (as Na₂CO₃): ≤ 0.5
Chloride (Cl): ≤ 0.004 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.003 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service