Skip to Content
Merck
All Photos(1)

Documents

D-033

Supelco

Dihydromorphine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3
CAS Number:
Molecular Weight:
287.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H](CC1)[C@@]2([H])[C@@]3([C@]1([H])[C@H](N(C)CC3)C4)C5=C4C=CC(O)=C5O2

InChI

1S/C17H21NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2,4,10-11,13,16,19-20H,3,5-8H2,1H3/t10-,11+,13-,16-,17-/m0/s1

InChI key

IJVCSMSMFSCRME-KBQPJGBKSA-N

General description

Dihydromorphine is a semisynthetic opioid analgesic sold under the trade names Paramorfan or Paramorphan for treatment of pain. Dihydromorphine is also a primary active metabolite of the analgesic opioid drug dihydrocodeine. This Snap-N-Spike® Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing and pain prescription monitoring to pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Grauert et al.
Journal of medicinal chemistry, 40(18), 2922-2930 (1997-08-29)
We have synthesized a series of stereoisomeric 6,7-benzomorphan derivatives with modified N-substituents and determined their ability to antagonize the N-methyl-D-aspartate (NMDA) receptor-channel complex in vitro and in vivo. The ability of the compounds to displace [3H]-MK-801 from the channel site
G B Stefano et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 113(3), 369-373 (1996-03-01)
A previous report demonstrated the presence of the newly discovered opiate alkaloid selective and opioid peptide insensitive mu3 receptor in ganglia of several invertebrate- and one vertebrate species as well as in microglial cells that had egressed from these ganglia
T Sagara et al.
Bioorganic & medicinal chemistry, 4(12), 2151-2166 (1996-12-01)
For three-dimensional understanding of the mechanisms that control potency and selectivity of the ligand binding at the atomic level, we have analysed opioid receptor-ligand interaction based on the receptor's 3D model. As a first step, we have constructed molecular models
M F Fromm et al.
Clinical pharmacology and therapeutics, 58(4), 374-382 (1995-10-01)
The opioid dihydrocodeine (DHC) is frequently used as an analgesic and antitussive agent. However, until now there have been no detailed data on dihydrocodeine metabolism in humans. We therefore investigated pathways that contribute to elimination of dihydrocodeine, and we tested
Peter A Crooks et al.
Bioorganic & medicinal chemistry letters, 16(16), 4291-4295 (2006-06-17)
A series of 3-O-acyl-6-O-sulfate esters of morphine, dihydromorphine, N-methylmorphinium iodide, codeine, and dihydrocodeine were prepared and evaluated for their ability to bind to mu-, delta-, kappa(1)-, kappa(2)-, and kappa(3)-opiate receptors. Several compounds exhibited good affinity for the mu-opiate receptor. Morphine-3-O-propionyl-6-O-sulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service