Skip to Content
Merck
All Photos(1)

Key Documents

S-001

Supelco

Secobarbital-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D5H13N2O3
CAS Number:
Molecular Weight:
243.31
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]\C([2H])=C(/[2H])C([2H])([2H])C1(C(C)CCC)C(=O)NC(=O)NC1=O

InChI

1S/C12H18N2O3/c1-4-6-8(3)12(7-5-2)9(15)13-11(17)14-10(12)16/h5,8H,2,4,6-7H2,1,3H3,(H2,13,14,15,16,17)/i2D2,5D,7D2

InChI key

KQPKPCNLIDLUMF-HHEIPOTRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for secobarbital testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, pain prescription monitoring, or urine drug testing. Secobarbital, sold under the trade name Seconal®, is a barbiturate used in the treatment of epilepsy, the short term treatment of insomnia and as a preoperative anesthetic.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Seconal is a registered trademark of Marathon Pharmaceuticals, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service