Skip to Content
Merck
All Photos(1)

Key Documents

M-046

Supelco

Morphine-6-β-D-glucuronide solution

1.0 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO9
CAS Number:
Molecular Weight:
461.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol: water (2:8)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)c35

InChI

1S/C23H27NO9/c1-24-7-6-23-10-3-5-13(31-22-17(28)15(26)16(27)19(33-22)21(29)30)20(23)32-18-12(25)4-2-9(14(18)23)8-11(10)24/h2-5,10-11,13,15-17,19-20,22,25-28H,6-8H2,1H3,(H,29,30)/t10-,11+,13-,15-,16-,17+,19-,20-,22+,23-/m0/s1

InChI key

GNJCUHZOSOYIEC-GAROZEBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified solution standard of Morphine-6-β-D-glucuronide at a concentration of 1.0 mg/mL for high volumes of sample testing. This new certified solution standard is suitable for LCMS or GCMS applications including forensic analysis or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Hannam et al.
Anaesthesia and intensive care, 40(5), 867-870 (2012-09-01)
A morphine plasma concentration/respiratory rate relationship has been described for both adults and children although that of its metabolite, morphine-6-glucuronide, remains uncertain. We describe this relationship in a child with end-stage renal failure who received repeat morphine administration over two
Claire Trécant et al.
European journal of medicinal chemistry, 46(9), 4035-4041 (2011-06-22)
Synthesis and biological evaluation of new derivatives of Morphine-6-Glucuronide (M6G) are described. M6G is an active metabolite of morphine which displays more analgesia than morphine with a superior side effect profile but with a less efficiently BBB penetration. These phenomena
Kristin Moksnes et al.
European journal of clinical pharmacology, 68(8), 1147-1156 (2012-03-01)
Our aim was to compare pharmacological aspects of two switching strategies from morphine/oxycodone to methadone; the stop and go (SAG) strategy in which methadone is started directly after the initial opioid has been stopped, and the 3-days switch (3DS), in
Toyofumi Suzuki
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 131(10), 1445-1451 (2011-10-04)
Opioid analgesics exhibit cationic properties under physiological conditions, and the mechanism underlying permeation of the blood-brain barrier thus cannot be fully explained by simple diffusion alone. Various types of transporters that exhibit substrate specificity are localized on the blood-brain barrier
Lin Yang et al.
The Journal of pharmacy and pharmacology, 65(4), 561-566 (2013-03-16)
The semi-synthetic bile salt, 12-oxochenodeoxycholate (OCDC also known as 12-monoketocholate), has been shown to enhance drug permeation across biological membranes with low cytotoxicity. Its effect on the analgesic potency and brain concentration of morphine 6-glucuronide (M6G) was studied in male

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service