Skip to Content
Merck
All Photos(1)

Key Documents

D-007

Supelco

Desmethyldoxepin solution

cis/trans, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO
CAS Number:
Molecular Weight:
265.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CNCC/C=C1C2=C(C=CC=C2)COC3=C/1C=CC=C3

InChI

1S/C18H19NO/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18/h2-5,7-11,19H,6,12-13H2,1H3/b16-10-

InChI key

HVKCEFHNSNZIHO-YBEGLDIGSA-N

General description

Desmethyldoxpein is the primary urinary metabolite of doxepin, a tricyclic antidepressant used for the treatment of sleep maintenance and sold under trade names such as Silenor® and Aponal. This analytical reference standard is suited for use in LC/MS or GC/MS methods for urine drug testing, clinical toxicology, and forensic analysis applications.

Application

Desmethyldoxepin solution may be used as an internal standard in the high performance liquid chromatographic (HPLC) determination of bupivacaine in samples of human serum.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Silenor is a registered trademark of Somaxon Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of bupivacaine in human serum
Lindberg.P.L.R
Journal of Chromatography. B, Biomedical Sciences and Applications, 309, 369-374 (1984)
Antonia Gronewold et al.
Forensic science international, 190(1-3), 74-79 (2009-06-16)
Body fluids and tissues in eight doxepin (Dox)-related deaths were investigated in order to prove whether the individual concentration of Dox, the concentration sum of parent drug and its active metabolite N-desmethyldoxepin (NDox) or the concentration ratio Dox/Ndox valuably contribute
Sebastian Härtter et al.
Pharmaceutical research, 19(7), 1034-1037 (2002-08-16)
This study was conducted to identify the cytochrome P450s (CYPs) responsible for the metabolism of the cis- and trans-isomers of the tricyclic antidepressant doxepin to its pharmacologically active N-desmethylmetabolite by in vitro techniques. The doxepin N-demethylation was studied by means
M Meyer-Barner et al.
European journal of clinical pharmacology, 58(4), 253-257 (2002-07-24)
Little information on the population pharmacokinetics of the tricyclic antidepressant doxepine and its pharmacologically active metabolite desmethyldoxepine is available. However, a more individualised drug therapy may be feasible if the influence of various patient characteristics on plasma concentration was known.
M Adamczyk et al.
Therapeutic drug monitoring, 17(4), 371-376 (1995-08-01)
A high-performance liquid chromatographic (HPLC) method was developed for the quantitative, simultaneous determination of the following four compounds in serum: E-doxepin, Z-doxepin, E-desmethyldoxepin, and Z-desmethyldoxepin. A 3-microns analytical silica column (6 x 100 mm) was employed with the mobile phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service