Skip to Content
Merck
All Photos(1)

Key Documents

C-909

Supelco

Clobazam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2O2
CAS Number:
Molecular Weight:
300.74
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN1C(=O)CC(=O)N(c2ccccc2)c3cc(Cl)ccc13

InChI

1S/C16H13ClN2O2/c1-18-13-8-7-11(17)9-14(13)19(16(21)10-15(18)20)12-5-3-2-4-6-12/h2-9H,10H2,1H3

InChI key

CXOXHMZGEKVPMT-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing. Clobazam, marketed under trade names Frisium, Urbanol, and Onfi® is benzodiazapene drug prescribed to treat anxiety and epilepsy.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Onfi is a registered trademark of Lundbeck LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New drugs 2013, Part I.
Daniel A Hussar
Nursing, 43(2), 36-46 (2013-01-11)
M Scott Perry et al.
Journal of child neurology, 28(1), 34-39 (2012-11-01)
We describe our experience using clobazam, examining efficacy for individual seizure types and analyzing for factors contributing to initial and sustained response. We retrospectively reviewed medical charts of children treated with clobazam. We collected patient and treatment characteristics and compared
Mark Walzer et al.
Pharmacotherapy, 32(4), 340-353 (2012-03-17)
To investigate potential drug-drug interactions between clobazam and cytochrome P450 (CYP) isoenzyme substrates, inhibitors, and inducers. Two, prospective, open-label, single-center, drug-drug interaction (DDI) studies and a population pharmacokinetics analysis of seven multicenter phase II-III trials. Clinical research unit. Fifty-four healthy
Marie Besson et al.
Basic & clinical pharmacology & toxicology, 112(3), 192-197 (2012-09-26)
Facilitation of spinal GABAergic inhibition with benzodiazepines (BZDs) reverses pain sensitization in animals; however, the use of BZDs in man is limited by their sedative effect. The antihyperalgesic effects of GABA(A) agonists are mediated by GABA(A) receptors containing α2 subunits
Dezhi Cao et al.
Epilepsia, 53(7), 1140-1145 (2012-05-15)
We previously reported a mutant mouse carrying a severe myoclonic epilepsy in infancy (SMEI) mutation in Scn1a. In this study, we examined the susceptibility to hyperthermia-induced seizures of heterozygous Scn1a mutant mice (Scn1a(RX/+)) and wild-type (Scn1a(+/+) ) mice. Then we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service