Skip to Content
Merck
All Photos(1)

Key Documents

C-033

Supelco

Clonidine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Cl2N3
CAS Number:
Molecular Weight:
230.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=CC(Cl)=C1NC2=NCCN2

InChI

1S/C9H9Cl2N3/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9/h1-3H,4-5H2,(H2,12,13,14)

InChI key

GJSURZIOUXUGAL-UHFFFAOYSA-N

Gene Information

General description

Clonidine is a pharmaceutical drug used to treat a wide variety of conditions including hypertension, ADHD, insomnia, chemical withdrawal, and anxiety. Clonidine is often prescribed in combination with other drugs, depending on the clinical condition. This analytical reference standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoher M Naja et al.
Anesthesia and analgesia, 117(1), 252-258 (2013-05-02)
Paravertebral blocks (PVBs) have been introduced as an alternative to general anesthesia for breast cancer surgeries. The addition of clonidine as an adjuvant in PVBs may enhance quality and duration of analgesia and significantly reduce the consumption of analgesics after
Conflicting information from TGA versus FDA may undermine compliance with use of medication.
Klaus Martin Beckmann
The Medical journal of Australia, 199(1), 28-29 (2013-07-09)
Janet E Wielenga-Boiten et al.
Journal of rehabilitation research and development, 49(9), 1349-1354 (2013-02-15)
Akathisia is reported to be one of the most common and disabling side effects of antipsychotics and other drugs. Akathisia is also a rare cause of psychomotor agitation in patients with traumatic brain injury (TBI). In this clinical note, we
I Davagnanam et al.
Eye (London, England), 27(3), 291-298 (2013-02-02)
The diagnosis of Horner's syndrome (HS) can be difficult, as patients rarely present with the classic triad of ptosis, miosis, and anhydrosis. Frequently, there are no associated symptoms to help determine or localise the underlying pathology. The onset of anisocoria
Shahram Borjian Boroojeny et al.
JPMA. The Journal of the Pakistan Medical Association, 62(12), 1285-1288 (2013-07-23)
To evaluate the efficacy of preoperative oral clonidine (5 g/kg) in preventing ocular hypertension in the early period after cataract surgery with posterior chamber intraocular lens implantation under general anaesthesia. This was a randomized double-blind clinical trial comprising of 62

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service