Skip to Content
Merck
All Photos(1)

Key Documents

880120P

Avanti

18:0 PEG2000 PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (880120P-1g)
pkg of 1 × 200 mg (880120P-200mg)
pkg of 1 × 25 mg (880120P-25mg)
pkg of 1 × 500 mg (880120P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C133H264NO55P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-131(135)186-128-130(189-132(136)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)129-188-190(138,139)187-37-36-134-133(137)185-127-126-184-125-124-183-123-122-182-121-120-181-119-118-180-1

InChI key

QSWJINHFEMDVEM-OJCMNYADSA-N

General description

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000 (18:0 PEG2000 PE) is conjugate, covalently attached to polyethylene glycol (PEG2000). It forms a stable micelle of 180kDa with phosphoethanolamine as hydrophobic core and hydrophilic PEG2000 as outer micelle surface.

Application

18:0 PEG2000 PE may be used in the nickel resin functionalization of quantum dots. It may also be used in liposome preparation.

Biochem/physiol Actions

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000 (18:0 PEG2000 PE) is an effective drug carrier and is biocompatible. PEG stabilizes and protects the micelle from phagocytic system. The use of PEG as a conjugate with phospholipids enables improved liposome preparation and effective drug encapsulation.

Packaging

20 mL Clear Glass Screw Cap Vial (880120P-1g)
20 mL Clear Glass Screw Cap Vial (880120P-500mg)
5 mL Amber Glass Screw Cap Vial (880120P-200mg)
5 mL Amber Glass Screw Cap Vial (880120P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relationship between targeting efficacy of liposomes and the dosage of targeting antibody using surface plasmon resonance
Xiang Y, et al.
Langmuir, 31(44), 12177-12186 (2015)
Inertial cavitation to non-invasively trigger and monitor intratumoral release of drug from intravenously delivered liposomes
Graham SM, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 178, 101-107 (2014)
Structure and dynamics of highly PEG-ylated sterically stabilized micelles in aqueous media
Vukovic L, et al.
Journal of the American Chemical Society, 133(34), 13481-13488 (2011)
P A Melnikov et al.
Bulletin of experimental biology and medicine (2019-11-24)
We studied the effect of IL-1β on the permeability of brain capillaries in healthy mice. Intravital microscopy demonstrated that parenteral administration of IL-1β was followed by an increase in vascular permeability ensuring passage of free Alexa488 fluorescent label through the
Ling Ye et al.
Nanotheranostics, 3(1), 54-65 (2019-01-22)
Rationale: Despite growing use of engineered nanomaterials (ENM) in applications from electronics to medicine, the potential risk to human health remains a critical concern within clinical use. ENM exposure during pregnancy can potentially cause reproductive toxicity even at levels that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service