Skip to Content
Merck
All Photos(2)

Key Documents

840875P

Avanti

18:1 PA

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphate (sodium salt); DOPA; PA(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8PNa
CAS Number:
Molecular Weight:
722.95
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840875P-1g)
pkg of 2 × 100 mg (840875P-200mg)
pkg of 5 × 100 mg (840875P-500mg)
pkg of 1 × 25 mg (840875P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C39H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,37H,3-16,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b19-17-,20-18-;/t37-;/m1./s1

InChI key

JUKKLZHXFHPJSF-ZBFGHDQJSA-M

General description

18:1 PA (1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt)) is an anionic lipid.

Application

18:1 PA (1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt)):
  • may be used in liposomes for liposome leakage assay
  • may be used to create liposome template to synthesize calcium phosphate (CaP) nanoshells
  • may be used as a component in liposomes/lipid bilayers
  • has been used an additive in the medium of neuron-glia mixed culture to study the effects of astrocyte phospholipase D1 (PLD1) on dendritic branching
  • has been used in chemiluminescent assay of cytochrome c Trp59

Biochem/physiol Actions

Phosphatidic acid (PA) acts as a lipid messenger.

Packaging

5 mL Clear Glass Sealed Ampule (840875P-200mg)
5 mL Clear Glass Sealed Ampule (840875P-25mg)
5 mL Clear Glass Sealed Ampule (840875P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840875P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of DOPA and DPPA liposome templates for the synthesis of calcium phosphate nanoshells
Yeo CH, et al.
Ceramics International, 38(1), 561-570 null
Shutao Guo et al.
Journal of controlled release : official journal of the Controlled Release Society, 174, 137-142 (2013-11-28)
The poor solubility of cisplatin (CDDP) often presents a major obstacle in the formulation of CDDP in nanoparticles (NPs) by traditional methods. We have developed a novel method for synthesizing CDDP NPs taking advantage of its poor solubility. By mixing
Xinhui Sun et al.
Cell cycle (Georgetown, Tex.), 14(5), 721-731 (2015-01-16)
It had been known for decades that primordial follicles in mammalian ovaries are assembled with definite numbers and represent the ovarian reserve throughout the reproductive life. Intra-oocyte PI3K/mTOR pathways have been indicated to play a central role on the activation
Zhenping Cao et al.
Nature communications, 10(1), 5783-5783 (2019-12-21)
The gut microbiota represents a huge community of microorganisms that play essential roles in immune modulation and homeostasis maintenance. Microbiota transplantation is an important approach to prevent and treat disease as it can inhibit pathogen colonization and positively modulate bacterial
Shireesha Sankella et al.
The Journal of biological chemistry, 289(8), 4762-4777 (2014-01-16)
In this study we examined the role of phosphatidic acid (PA) in hepatic glucose production (HGP) and development of hepatic insulin resistance in mice that lack 1-acylglycerol-3-phosphate O-acyltransferase 2 (AGPAT2). Liver lysophosphatidic acid and PA levels were increased ∼2- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service