Skip to Content
Merck
All Photos(2)

Key Documents

850365P

Avanti

18:0 PC (DSPC)

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioctadecanoyl-sn-glycero-3-phosphocholine; DSPC; PC(18:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H88NO8P
CAS Number:
Molecular Weight:
790.15
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-distearoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850365P-1G)
pkg of 1 × 200 mg (850365P-200MG)
pkg of 1 × 25 mg (850365P-25MG)
pkg of 1 × 500 mg (850365P-500MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1

InChI key

NRJAVPSFFCBXDT-HUESYALOSA-N

General description

1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC) is a phospholtidylcholine with alkyl chain comprising 18 carbons. It has phophatidylcholine head group and its structure is similar to 1,2-dipalmitoyl-sn-glycero-3-phosphatidylcholine (DPPC).
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:0 PC (DSPC) has been used:
  • in the microbubble monolayer preparation for biomolecule delivery studies into protoplasts
  • in the preparation of liposomes for immobilization studies
  • in the generation of porphyrin-phospholipid (PoP) liposomes for irinotecan (IRT) and doxorubicin delivery

Biochem/physiol Actions

1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC) is used in combination with other members of phosphatidylcholine for the generation of mixed bilayer system. It is majorly used in conventional and stealth liposome preparation for drug delivery studies.

Packaging

20 mL Clear Glass Screw Cap Vial (850365P-1G)
20 mL Clear Glass Screw Cap Vial (850365P-500MG)
5 mL Amber Glass Screw Cap Vial (850365P-200MG)
5 mL Amber Glass Screw Cap Vial (850365P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of PEGylated and conventional liposomes as carriers for shikonin
Tsermentseli S, et al.
Fluids, 3(2), 36-36 (2018)
Investigating the structure of multicomponent gel-phase lipid bilayers
Hartkamp R, et al.
Biophysical Journal, 111(4), 813-823 (2016)
Ana Reis et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183133-183133 (2019-12-01)
Food polyphenols in fruits juices, tea, coffee, wine and beer confer sensory properties such as colour, astringency and bitterness. The development of functional healthy drinks without the unpleasant sensory feeling is boosting research for a clearer understanding on the interactions
Viktoriya Dubrovskaya et al.
Immunity, 51(5), 915-929 (2019-11-17)
The elicitation of broadly neutralizing antibodies (bNAbs) against the HIV-1 envelope glycoprotein (Env) trimer remains a major vaccine challenge. Most cross-conserved protein determinants are occluded by self-N-glycan shielding, limiting B cell recognition of the underlying polypeptide surface. The exceptions to
Kenneth L Clark et al.
Molecular therapy. Nucleic acids, 2, e65-e65 (2013-01-17)
Lung pathology in cystic fibrosis is linked to dehydration of the airways epithelial surface which in part results from inappropriately raised sodium reabsorption through the epithelial sodium channel (ENaC). To identify a small-interfering RNA (siRNA) which selectively inhibits ENaC expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service