Skip to Content
Merck
All Photos(1)

Key Documents

W310107

Sigma-Aldrich

Valeric acid

≥99%, FCC, FG

Synonym(s):

n-Valeric acid, Pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
FEMA Number:
3101
Beilstein:
969454
EC Number:
Council of Europe no.:
7
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.007
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

707 °F

expl. lim.

7.6 %

refractive index

n20/D 1.408 (lit.)

bp

110-111 °C/10 mmHg (lit.)
185 °C (lit.)

mp

−20-−18 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cheesy; acidic; rancid

SMILES string

CCCCC(O)=O

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Valeric acid is one of the volatile flavor compounds identified in soy sauce, roasted barley, Auricularia polytricha essential oil and rice.

Application


  • Synthesis and biodegradation testing of some synthetic oils based on ester.: Examines the biodegradability of ester-based synthetic oils, with valeric acid esters playing a crucial role in environmental sustainability of industrial lubricants (Shazly RIE et al., 2024).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of Volatile Odor Components of Superheated Steam-cooked Rice with a Less Stale Flavor.
Takemitsu H, et al.
Food Science and Technology Research, 22(6), 771-778 (2016)
Volatile flavor components of Korean Auricularia polytricha (Mont.) Sacc. mushroom.
Lee JG, et al.
Agricultural and Biological Chemistry, 38(6), 546-548 (1995)
Studies on Flavor Components of Roasted Barley.
Wang PS, et al.
Agricultural and Biological Chemistry, 34(4), 561-567 (1970)
Flavor and taste compounds analysis in Chinese solid fermented soy sauce.
Yanfang Z & Wenyi T.
African Journal of Biotechnology, 8(4) (2009)
M F Böttcher et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 30(11), 1590-1596 (2000-11-09)
The prevalence of allergic diseases has increased particularly over the past 30-40 years. A reduced microbial stimulation during infancy may result in a development of a disturbed balance between Th1- and Th2-like immunity. The gut flora is, quantitatively, the most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service