Skip to Content
Merck
All Photos(1)

Key Documents

W292400

Sigma-Aldrich

Propionic acid

≥99.5%, FCC, FG

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
FEMA Number:
2924
Beilstein:
506071
EC Number:
Council of Europe no.:
3
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39021305
PubChem Substance ID:
Flavis number:
8.003
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 178.1010
FDA 21 CFR 184.1081

vapor density

2.55 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

Assay

≥99.5%

autoignition temp.

955 °F

expl. lim.

12.1 %

refractive index

n20/D 1.386 (lit.)

bp

141 °C (lit.)

mp

−24-−23 °C (lit.)

solubility

H2O: soluble

density

0.993 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

dairy; pungent; acidic

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionic acid (PA) is a naturally occurring carboxylic acid, which in its pure state exists as a colorless corrosive liquid with an unpleasant odor. Propionic acid is an excellent raw material as it is stable, cheap and safe and can even be used as a food additive.
Propionic acid is a volatile fatty acid (VFA) formed in the rumen of ruminants by bacterial fermentations.

Application


  • Distribution Characteristics of Low-Molecular-Weight Organic Acids in Reclaimed Soil Filled with Fly Ash: A Study.: This research investigates the distribution of low-molecular-weight organic acids, including propionic acid, in reclaimed soil, highlighting environmental and soil health implications (Zheng et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of propionic acid in animal tissues I. Enzymatic conversion of propionate to succinate.
Flavin M & Ochoa S.
The Journal of Biological Chemistry, 229(2), 965-979 (1957)
Interconversions and production of volatile fatty acids in the sheep rumen.
Bergman EN, et al.
The Biochemical Journal, 97(1), 53-58 (1965)
Biological effects of propionic acid in humans; metabolism, potential applications and underlying mechanisms
Sa'ad, H., et al.
Molecular and Cellular Biology, 1801 (11), 1175-1183 (2010)
Nucleophilic ?-Carbon Activation of Propionic Acid as a 3-Carbon Synthon by Carbene Organocatalysis
Jin, Zhichao, et al.
Chemistry?A European Journal , 21 (26), 9360-9363 (2015)
Flour and Breads and their Fortification in Health and Disease Prevention.
Victor R. Preedy, Ronald Ross Watson, Vinood B
Technology & Engineering, 476-476 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service