Skip to Content
Merck
All Photos(1)

Key Documents

W206105

Sigma-Aldrich

α-Amylcinnamaldehyde

mixture of cis and trans, ≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C[(CH2)4CH3]CHO
CAS Number:
Molecular Weight:
202.29
FEMA Number:
2061
Council of Europe no.:
128
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.040
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

contains

BHT as stabilizer

composition

contains IFRA and EU 1223/2009 restricted α-amylcinnamaldehyde

refractive index

n20/D 1.557 (lit.)

bp

287-290 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

amyl cinnamal

Organoleptic

fruity; floral; sweet

SMILES string

[H]C(/C(CCCCC)=C/C1=CC=CC=C1)=O

InChI

1S/C14H18O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11-12H,2-3,5,10H2,1H3/b14-11+

InChI key

HMKKIXGYKWDQSV-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eiram N Elahi et al.
Chemical research in toxicology, 17(3), 301-310 (2004-03-18)
Skin protein modification (haptenation) is thought to be a key step in the manifestation of sensitization to low molecular mass chemicals (<500 g/mol). For sensitizing chemicals that are not protein reactive, it is hypothesised that metabolic activation can convert such
J D Guin et al.
Journal of the American Academy of Dermatology, 8(1), 76-80 (1983-01-01)
Sensitivity to alpha-amylcinnamic aldehyde (alpha-AcAld) is apparently uncommon, but, like allergy to alpha-amylcinnamic alcohol (alpha-AcAlc), it often accompanies allergy to the perfume in Mycolog cream. Although alpha-AcAlc is a known ingredient, alpha-AcAld is not. However, gas-liquid chromatographic analysis shows alpha-AcAld
S C Rastogi et al.
Contact dermatitis, 38(1), 29-35 (1998-03-21)
Deodorants are one of the most frequently used types of cosmetics and side-effects from them are common. Recent studies relate perfume allergy to this type of product. 73 deodorants were analyzed by gas chromatography--mass spectrometry for the determination of the
Rowe, D.
Perfumer & Flavorist, 30, 20-20 (2005)
S C Rastogi et al.
Contact dermatitis, 34(6), 423-426 (1996-06-01)
In the present study, we have investigated 42 cosmetic products based on natural ingredients for content of 11 fragrance substances: geraniol, hydroxycitronellal, eugenol, isoeugenol, cinnamic aldehyde, cinnamic alcohol, alpha-amylcinnamic aldehyde, citral, coumarin, dihydrocoumarin and alpha-hexylcinnamic aldehyde. The study revealed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service