Skip to Content
Merck
All Photos(1)

Key Documents

W237809

Sigma-Aldrich

Diethyl L-tartrate

≥99%, FG

Synonym(s):

(+)-Diethyl L-tartrate, L-(+)-Tartaric acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CO2C2H5]2
CAS Number:
Molecular Weight:
206.19
FEMA Number:
2378
Beilstein:
1727145
EC Number:
Council of Europe no.:
440
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.446
NACRES:
NA.21
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥99%

optical activity

[α]20/D +8.5°, neat

refractive index

n20/D 1.446 (lit.)

bp

280 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; wine-like

SMILES string

CCOC(=O)[C@H](O)[C@@H](O)C(=O)OCC

InChI

1S/C8H14O6/c1-3-13-7(11)5(9)6(10)8(12)14-4-2/h5-6,9-10H,3-4H2,1-2H3/t5-,6-/m1/s1

InChI key

YSAVZVORKRDODB-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis of l-threitol-based crown ethers and their application as enantioselective phase transfer catalyst in Michael additions.: This study synthesizes l-threitol-based crown ethers using diethyl ʟ-tartrate and explores their efficacy as enantioselective phase transfer catalysts in Michael additions, highlighting their potential in asymmetric synthesis (Rapi et al., 2017).

  • A facile approach for the synthesis of C13-C24 fragments of maltepolides A, C and D.: This research demonstrates a novel synthesis method for C13-C24 fragments of maltepolides A, C, and D using diethyl ʟ-tartrate, facilitating the study and development of these bioactive compounds (Rao & Srihari, 2016).

  • Development of diacyltetrol lipids as activators for the C1 domain of protein kinase C.: This research introduces diacyltetrol lipids synthesized from diethyl ʟ-tartrate, which act as activators for the C1 domain of protein kinase C, offering insights into signal transduction and therapeutic applications (Mamidi et al., 2012).

  • Total synthesis of broussonetine F: the orthoamide Overman rearrangement of an allylic diol.: The paper presents the total synthesis of broussonetine F, utilizing diethyl ʟ-tartrate in an orthoamide Overman rearrangement, showcasing a novel synthetic route for complex natural products (Hama et al., 2011).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly A Kahle et al.
Electrophoresis, 28(15), 2644-2657 (2007-06-29)
In this second study on dual-chirality microemulsions containing a chiral surfactant and a chiral oil, a less hydrophobic and lower interfacial tension chiral oil, diethyl tartrate, is employed (Part 1, Foley, J. P. et al.., Electrophoresis, DOI: 10.1002/elps.200600551). Six stereochemical
Total synthesis of the light-harvesting carotenoid peridinin.
Thomas Olpp et al.
Angewandte Chemie (International ed. in English), 45(24), 4023-4027 (2006-05-10)
Jiang Weng et al.
The Journal of organic chemistry, 75(9), 3125-3128 (2010-04-17)
A short and practical synthesis of oseltamivir was accomplished in 11 steps from inexpensive and abundant diethyl D-tartrate starting material. This azide-free route featured an asymmetric aza-Henry reaction and a domino nitro-Michael/Horner-Wadsworth-Emmons (HWE) reaction as the key steps to construct
Aman Ullah et al.
Biomacromolecules, 12(10), 3826-3832 (2011-09-06)
Poultry feather quills have been extruded in a twin screw extruder with sodium sulfite treatment as a reducing agent. The effect of four different plasticizers (ethylene glycol, propylene glycol, glycerol, and diethyl tartrate) on the thermoplastic properties was then investigated.
Naoki Miyakoshi et al.
The Journal of organic chemistry, 70(15), 6045-6052 (2005-07-16)
A highly stereoselective method for constructing a (2E)-methoxymethylidene-1,6-dioxaspiro[4.5]decane skeleton has been developed on the basis of the palladium(II)-catalyzed ring-closing reaction of the 3,4-dioxygenated-9-hydroxy-1-nonyn-5-one derivatives as a crucial step. The newly developed procedures could be successfully applied to the first total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service