227803
(−)-Diisopropyl D-tartrate
98%
Synonym(s):
D-(−)-Tartaric acid diisopropyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[-CH(OH)CO2CH(CH3)2]2
CAS Number:
Molecular Weight:
234.25
Beilstein:
5265431
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
optical activity
[α]23/D −17°, neat
optical purity
ee: ≥99% (GLC)
refractive index
n20/D 1.439 (lit.)
density
1.119 g/mL at 20 °C (lit.)
functional group
ester
hydroxyl
SMILES string
CC(C)OC(=O)[C@@H](O)[C@H](O)C(=O)OC(C)C
InChI
1S/C10H18O6/c1-5(2)15-9(13)7(11)8(12)10(14)16-6(3)4/h5-8,11-12H,1-4H3/t7-,8-/m0/s1
InChI key
XEBCWEDRGPSHQH-YUMQZZPRSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Both D- and L-forms are reagents for kinetic resolution of racemic allylic alcohols and α-furfuryl amides by enantioselective epoxidation.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrahedron, 49, 2641-2641 (1993)
Aldrichimica Acta, 18, 53-53 (1985)
The Journal of Organic Chemistry, 59, 1444-1444 (1994)
Journal of the American Chemical Society, 115, 12208-12208 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service