Skip to Content
Merck
All Photos(2)

Key Documents

T38091

Sigma-Aldrich

m-Tolylacetic acid

99%

Synonym(s):

3-Methylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2CO2H
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

64-66 °C (lit.)

SMILES string

Cc1cccc(CC(O)=O)c1

InChI

1S/C9H10O2/c1-7-3-2-4-8(5-7)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

GJMPSRSMBJLKKB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Nomeir et al.
Analytical biochemistry, 135(2), 296-303 (1983-12-01)
A method utilizing high-performance liquid chromatography (HPLC) has been developed for the analysis of tri-o-cresyl phosphate (TOCP) and its possible metabolites, o-cresyl dihydrogen phosphate, di-o-cresyl hydrogen phosphate, o-hydroxybenzyl alcohol, o-cresol, saligenin cyclic-o-tolyl phosphate [2-(o-cresyl)-4H-1:3:2:benzodioxaphosphoran-2-one], salicylic acid, salicylaldehyde, hydroxymethyl TOCP (di-o-cresyl
[Further studies on the metabolic behavior of some aromatic and hydroaromatic mono- and dicarboxylic acids].
K BERNHARD et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 310(1-2), 37-43 (1958-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service