Skip to Content
Merck
All Photos(4)

Key Documents

M19201

Sigma-Aldrich

4-Methoxyphenylacetic acid

ReagentPlus®, 99%

Synonym(s):

Homoanisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

bp

140 °C/3 mmHg (lit.)

mp

84-86 °C (lit.)

SMILES string

COc1ccc(CC(O)=O)cc1

InChI

1S/C9H10O3/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

NRPFNQUDKRYCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Methoxyphenylacetic acid can be used:
  • To prepare methyl 4-methoxyphenylacetate by esterification with dimethyl carbonate using mesoporous sulfated zirconia catalyst.
  • As a ligand to synthesize pharmacologically important dinuclear gallium(III) and phenyltin(IV) carboxylate metal complexes.
  • As a reactant to synthesize hydroxylated (E)-stilbenes by reacting with substituted benzaldehydes via Perkin reaction.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

379.4 °F

Flash Point(C)

193 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esterification of 4-methoxyphenylacetic acid with dimethyl carbonate over mesoporous sulfated zirconia
Devulapelli VG and Weng H-S
Catalysis Communications, 10(13), 1711-1717 (2009)
An unusual, mild and convenient one-pot two-step access to (E)-stilbenes from hydroxy-substituted benzaldehydes and phenylacetic acids under microwave activation: a new facet of the classical Perkin reaction
Sinha AK, et al.
Tetrahedron, 63(45), 11070-11077 (2007)
Santiago Gómez-Ruiz et al.
Journal of inorganic biochemistry, 102(12), 2087-2096 (2008-09-02)
The reaction of 3-methoxyphenylacetic acid (3-MPAH), 4-methoxyphenylacetic acid (4-MPAH), 2,5-dimethyl-3-furoic acid (DMFUH) or 1,4-benzodioxane-6-carboxylic acid (BZDOH) with triphenyltin(IV) chloride (1:1) or diphenyltin(IV) dichloride (2:1) in the presence of triethylamine yielded the compounds [SnPh3(3-MPA)] (1), [SnPh3(4-MPA)] (2), [SnPh3(DMFU)] (3), [SnPh3(BZDO)] (4)
Milena R Kaluderović et al.
European journal of medicinal chemistry, 45(2), 519-525 (2009-11-21)
The reaction of 3-methoxyphenylacetic acid, 4-methoxyphenylacetic acid, mesitylthioacetic acid, 2,5-dimethyl-3-furoic acid and 1,4-benzodioxane-6-carboxylic acid with trimethylgallium (1:1) yielded the dimeric complexes [Me(2)Ga(micro-O(2)CCH(2)C(6)H(4)-3-OMe)](2) (1), [Me(2)Ga(micro-O(2)CCH(2)C(6)H(4)-4-OMe)](2) (2), [Me(2)Ga(micro-O(2)CCH(2)SMes)](2) (3) (Mes=2,4,6-Me(3)C(6)H(2)), [Me(2)Ga{micro-O(2)C(Fur)}](2) (4) (Fur=2,5-dimethylfuran) and [Me(2)Ga{micro-O(2)C(Bdo)}](2) (5) (Bdo=1,4-benzodioxane) respectively. The molecular structure of
Ki Hyun Kim et al.
Bioorganic & medicinal chemistry letters, 21(8), 2270-2273 (2011-03-23)
A bioassay-guided fractionation and chemical investigation of the trunk of Berberis koreana resulted in the isolation and identification of a new sesquilignan, named berbikonol (1), along with fourteen known lignan derivatives (2-15) and a new phenolic compound, named berfussinol (16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service