Skip to Content
Merck
All Photos(1)

Documents

103608

Sigma-Aldrich

3-Nitrophenylacetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
2050088
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

117-120 °C (lit.)

SMILES string

OC(=O)Cc1cccc(c1)[N+]([O-])=O

InChI

1S/C8H7NO4/c10-8(11)5-6-2-1-3-7(4-6)9(12)13/h1-4H,5H2,(H,10,11)

InChI key

WUKHOVCMWXMOOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrophenylacetic acid is obtained by mild oxidation of 4-amino phenylacetic acid using potassium peroxymonosulfate as oxidizing agent in acetone.

Application

3-Nitrophenylacetic acid was used to study photodecarboxylation of nitrophenylacetate ions in aqueous solution. 3-Nitrophenylacetic acid was used in study to synthesize an azobenzene amino acid, having potent use as photo-inducible conformational switch in polypeptides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mild oxidation of aromatic amines.
Webb KS and Seneviratne S.
Tetrahedron Letters, 36(14), 2377-2378 (1995)
Photodecarboxylation of nitrophenylacetate ions.
Margerum JD and Petrusis CT.
Journal of the American Chemical Society, 91(10), 2467-2472 (1969)
Andreas Aemissegger et al.
Nature protocols, 2(1), 161-167 (2007-04-03)
The synthesis of an azobenzene amino acid (aa) for use as a photo-inducible conformational switch in polypeptides is described. The compound can be easily incorporated into an aa sequence by solid-phase peptide synthesis using standard 9-fluorenylmethoxycarbonyl methods. A reversible conformational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service