Skip to Content
Merck
All Photos(1)

Key Documents

H6401

Sigma-Aldrich

Hexachloropropene

≥90%

Synonym(s):

Perchloropropene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3CCl=CCl2
CAS Number:
Molecular Weight:
248.75
Beilstein:
1766640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.3 (vs air)

Quality Level

Assay

≥90%

form

liquid

refractive index

n20/D 1.549 (lit.)

bp

209-210 °C (lit.)

density

1.765 g/mL at 25 °C (lit.)

SMILES string

Cl\C(Cl)=C(\Cl)C(Cl)(Cl)Cl

InChI

1S/C3Cl6/c4-1(2(5)6)3(7,8)9

InChI key

VFDYKPARTDCDCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Neudecker et al.
Mutation research, 170(1-2), 1-9 (1986-04-01)
In the presence of S9 mix all allylic chloropropenes tested exert considerable indirect mutagenic activity which is most pronounced for 1,2,3-trichloropropene. Lower as well as higher chlorinated derivatives are clearly less mutagenic. Longer than standard incubation time (120 min instead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service