H2120
Heptaldehyde
95%
Synonym(s):
1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)5CHO
CAS Number:
Molecular Weight:
114.19
Beilstein:
1560236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.413 (lit.)
bp
153 °C (lit.)
mp
−43 °C (lit.)
density
0.817 g/mL at 25 °C (lit.)
SMILES string
CCCCCCC=O
InChI
1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3
InChI key
FXHGMKSSBGDXIY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F - closed cup
Flash Point(C)
35 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Romina B Barrozo et al.
The European journal of neuroscience, 33(10), 1841-1850 (2011-04-15)
Innate behaviours in animals can be influenced by several factors, such as the environment, experience, or physiological status. This behavioural plasticity originates from changes in the underlying neuronal substrate. A well-described form of plasticity is induced by mating. In both
Hui Xu et al.
Journal of chromatography. A, 1217(16), 2371-2375 (2009-10-13)
In this paper, an ultrasound-assisted headspace liquid-phase microextraction with in-drop derivatization was developed for the extraction and determination of hexanal and heptanal as the biomarkers in human blood. In the method, a polychloroprene rubber (PCR) tube was utilized as container
Carla Cioffi et al.
Chemical communications (Cambridge, England), (20)(20), 2129-2131 (2006-05-17)
The functionalisation of single wall carbon nanohorns via 1,3-dipolar cycloaddition has been achieved, and the products have been characterised by spectroscopy, microscopy and thermogravimetry.
Hui-Juan Zhang et al.
Journal of chromatography. A, 1160(1-2), 114-119 (2007-05-01)
A simple, rapid and sensitive method for the determination of hexanal and heptanal in plasma by high-performance liquid chromatography (HPLC) has been developed, which is based on polymer monolith microextraction (PMME) with in situ derivatization. 2,4-dinitrophenylhydrazine (DNPH) as a derivatizing
Nina Deisig et al.
PloS one, 7(3), e33159-e33159 (2012-03-20)
Most animals rely on olfaction to find sexual partners, food or a habitat. The olfactory system faces the challenge of extracting meaningful information from a noisy odorous environment. In most moth species, males respond to sex pheromone emitted by females
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service