Skip to Content
Merck
All Photos(3)

Key Documents

W278220

Sigma-Aldrich

Nonanal

greener alternative

natural, ≥98%, FG

Synonym(s):

Aldehyde C9, Nonyl aldehyde, Pelargonaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24
FEMA Number:
2782
Beilstein:
1236701
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.025
NACRES:
NA.21

biological source

botanical

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor pressure

~0.26 mmHg ( 25 °C)

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.424 (lit.)

bp

93 °C/23 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fatty; fresh; green; waxy; citrus

SMILES string

CCCCCCCCC=O

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3

InChI key

GYHFUZHODSMOHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonanal is a volatile organic compound with sweet orange and rose flavor. It is found in the essential oils of cinnamon, lemongrass, milkweed, rose, pine, and citrus oil.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application

Nonanal can be used as a flavoring agent in the food, and cosmetic industries. It is also used to improve rose-like odor in a variety of perfumery products.

Other Notes

Natural occurrence: Apple, avocado, balck currants, cooked beef, cucumber, fish, grapefruit, lemon, lime, mandarin, orange and peach.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olfactory Stimulation Effect of Aldehydes, Nonanal, and Decanal on the Human Electroencephalographic Activity, According to Nostril Variation
Kim Minju, et al.
Biomedicines, 7(3), 57-57 (2019)
Antidiarrhoeal activity of nonanal, an aldehyde isolated from Artemisia ludoviciana
Zavala-Sanchez MA, et al.
Pharmaceutical biology, 40(4), 263-268 (2002)
Nonanal Stimulates Growth Factors via Cyclic Adenosine Monophosphate (cAMP) Signaling in Human Hair Follicle Dermal Papilla Cells
Park Soyoon, et al.
International Journal of Molecular Sciences, 21(21), 8054-8054 (2020)
Investigation on the utilization possibility of orange (Citrus sinensis var. Valencia) oil extracted by microwave pretreatment-improved steam distillation as natural flavoring agent based on its characteristics analysis
Shaghaleh H, et al.
Journal of Essential Oil-Bearing Plants, 21(2), 298-316 (2018)
Asplenioideae species as a reservoir of volatile organic compounds with potential therapeutic properties
Froissard D, et al.
Natural Product Communications, 10(6) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service