Skip to Content
Merck
All Photos(1)

Key Documents

324140

Sigma-Aldrich

trans-2-Heptenal

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCHO
CAS Number:
Molecular Weight:
112.17
Beilstein:
1700822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

≥95%

form

liquid

refractive index

n20/D 1.450 (lit.)

bp

90-91 °C/50 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)\C([H])=C(/[H])CCCC

InChI

1S/C7H12O/c1-2-3-4-5-6-7-8/h5-7H,2-4H2,1H3/b6-5+

InChI key

NDFKTBCGKNOHPJ-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte L Steffensen et al.
Journal of agricultural and food chemistry, 50(25), 7392-7395 (2002-11-28)
In the present study, the aldehyde-induced pro-oxidative activity of xanthine oxidase was followed in an accelerated raw milk system using spin-trap electron spin resonance (ESR) spectroscopy. The aldehydes acetaldehyde, propanal, hexanal, trans-2-hexenal, trans-2-heptenal, trans-2-nonenal, and 3-methyl-2-butenal were all found to
M E Davis et al.
Physical chemistry chemical physics : PCCP, 9(18), 2240-2248 (2007-05-10)
Rate coefficients for the gas-phase reaction of the OH radical with (E)-2-pentenal (CH(3)CH(2)CH[double bond]CHCHO), (E)-2-hexenal (CH(3)(CH(2))(2)CH[double bond]CHCHO), and (E)-2-heptenal (CH(3)(CH(2))(3)CH[double bond]CHCHO), a series of unsaturated aldehydes, over the temperature range 244-374 K at pressures between 23 and 150 Torr (He
R Canonero et al.
Mutation research, 244(2), 153-156 (1990-06-01)
Five alk-2-enals--pent-2-enal, hex-2-enal, hept-2-enal, oct-2-enal and non-2-enal--produced by lipid peroxidation were tested for mutagenic activity in V79 Chinese hamster cells. At concentrations ranging from 0.003 to 0.3 mM all 5 alk-2-enals induced a dose-dependent increase in the frequency of 6-thioguanine-resistant
Richard Gminski et al.
Toxicology letters, 196(1), 33-41 (2010-04-07)
Due to the massive reduction of air-change rates in modern, energy-saving houses and dwellings, the contribution of volatile organic compound (VOCs) emissions from wood-based materials to indoor air quality has become increasingly important. To evaluate toxicity of VOC mixtures typically
G H Lee et al.
Journal of food science, 73(6), C500-C505 (2009-02-26)
The fruit of Lycii fructus has been used as a tonic medicine and a long-term healthy food without side effect in Asia. An increase in the demand for natural healthy food, L. fructus has been thought as a source of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service