Skip to Content
Merck
All Photos(2)

Key Documents

GF09275823

Beryllium

wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%

Synonym(s):

Beryllium, BE005110, Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
MDL number:
UNSPSC Code:
12141501
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.7%

form

wire

autoignition temp.

1198 °F

manufacturer/tradename

Goodfellow 092-758-23

resistivity

4.46 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Harper
Journal of environmental monitoring : JEM, 8(6), 598-604 (2006-06-13)
Standardized conventions governing the fractions of airborne particles that can penetrate the human head airways, the thoracic airways and the alveolar spaces have been internationally (although not universally) adopted. Several agencies involved in setting limit values for occupational exposure concentrations
Massimo Amicosante et al.
Clinical immunology (Orlando, Fla.), 121(2), 134-143 (2006-05-16)
Chronic beryllium disease (CBD) is a granulomatous lung disorder caused by beryllium exposure in the workplace and is characterized by the accumulation of beryllium-specific CD4(+) T cells. Depending on genetic susceptibility and the nature of the exposure, CBD occurs in
Paolo Boffetta et al.
Critical reviews in toxicology, 42(2), 107-118 (2012-01-27)
There is controversy on whether occupational exposure to beryllium causes lung cancer. We conducted a systematic review of epidemiologic studies on cancer among workers exposed to beryllium, including a study of seven U.S. production plants which has been recently updated
Shaodong Dai et al.
Current opinion in immunology, 25(6), 775-780 (2013-08-28)
Chronic beryllium disease (CBD) is a granulomatous lung disorder caused by a hypersensitivity to beryllium and characterized by the accumulation of beryllium-specific CD4(+) T cells in the lung. Genetic susceptibility to beryllium-induced disease is strongly associated with HLA-DP alleles possessing
L Mangin-Thro et al.
Nature communications, 6, 7705-7705 (2015-07-04)
The pseudo-gap phenomenon in copper oxide superconductors is central to any description of these materials as it prefigures the superconducting state itself. A magnetic intra-unit-cell order was found to occur just at the pseudo-gap temperature in four cuprate high-Tc superconducting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service