Product Description
Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%
Sigma-Aldrich
Beryllium, chunks, 99.9%
Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.05mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%
Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%
Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%
Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%
Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%