Skip to Content
Merck
Beryllium foil, thickness 2.0 mm, size 50 × 50 mm, purity 99%

Beryllium

Beryllium foil, thickness 2.0 mm, size 50 × 50 mm, purity 99%
Synonym(s):
Glucinium, Beryllium
Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Showing 1-20 of 46
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3