Skip to Content
Merck
All Photos(4)

Key Documents

A9891

Sigma-Aldrich

7-Amino-4-methylcoumarin

chromophore for enzyme substrates, fluorogenic, ≥98% (HPLC), powder

Synonym(s):

Coumarin 120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
142231
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

7-Amino-4-methylcoumarin, Chromophore for substrates

Quality Level

description

chromophore for enzyme substrates

Assay

≥98% (HPLC)

form

powder

mp

223-226 °C (lit.)

solubility

acetone: 10 mg/mL, clear, colorless to yellow

fluorescence

λex 365 nm; λem 440 nm in ethanol(lit.)

storage temp.

2-8°C

SMILES string

CC1=CC(=O)Oc2cc(N)ccc12

InChI

1S/C10H9NO2/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5H,11H2,1H3

InChI key

GLNDAGDHSLMOKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-Amino-4-methylcoumarin has been used:
  • to determine cathepsin B like- activity
  • as a substrate for leucine aminopeptidase (LAP)
  • in chymotryptic assay
  • as a standard to detect the activation of caspase-3 during
  • sanguinarine-induced damage
  • protection by human DEAD-box DDX3

Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and sialylated tri-and tetrasaccharides.

Biochem/physiol Actions

7-Amino-4-methylcoumarin is a coumarin derivative, which serves as a fluorescence reference standard to screen protease inhibitors.

Preparation Note

Soluble in DMSO (10 mg/mL), DMF, and acetone (10 mg/mL).

Other Notes

Reagent for preparing new fluorogenic AMC-based substrates for cystine aminopeptidase (and other hydrolases); used as reference compound in the enzyme assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Issues in Chemistry and General Chemical Research (2012)
Regulation of p21 expression for anti-apoptotic activity of DDX3 against sanguinarine-induced cell death on intrinsic pathway
Nguyen CN, et al.
Phytomedicine, 153096-153096 (2019)
Chymase uptake by cardiomyocytes results in myosin degradation in cardiac volume overload
Powell PC, et al.
Heliyon, 5(4), e01397-e01397 (2019)
Significant activities of extracellular enzymes from a brown tide in the coastal waters of Qinhuangdao, China
Ou L, et al.
Harmful Algae, 74, 1-9 (2018)
Sander S van Berkel et al.
ChemMedChem, 7(4), 606-617 (2012-02-02)
The synthesis of a series of peptides containing C-terminal 7-amino-4-methylcoumarin (AMC) for use in the thrombin generation test (TGT) is described. The lead structure in this project was H-Gly-Gly-Arg-AMC, of which the water solubility and kinetic parameters (K(M) and k(cat))

Articles

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service