All Photos(1)
About This Item
Linear Formula:
CH3CH2CHBrCH3
CAS Number:
Molecular Weight:
137.02
Beilstein:
505949
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.437 (lit.)
bp
91 °C (lit.)
density
1.255 g/mL at 25 °C (lit.)
SMILES string
CCC(C)Br
InChI
1S/C4H9Br/c1-3-4(2)5/h4H,3H2,1-2H3
InChI key
UPSXAPQYNGXVBF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-Bromobutane is an alkyl halide. Degradation of 2-bromobutane by reductive dehalogenation in the presence of nickel-aluminum alloy in potassium hydroxide solution has been reported. Gas chromatographic determination of 2-bromobutane in mixture of alkyl bromides has been reported. Stereochemistry of dehydrobromination of 2-bromobutane over KOH-silica has been investigated.
Viscosities of binary mixtures of 2-bromobutane and the isomeric forms of butanol have been evaluated at 298.15 and 313.15K. 2,2-dibromobutane, meso-2,3-dibromobutane and dl-2,3-dibromobutane are formed as major photobromination reaction products, during its reaction with molecular bromine. Smaller yields of 1,2-dibromobutane and 2,2,3-tribromobutane are also obtained in this reaction.
Application
2-Bromobutane may be used in the synthesis of series of non-nucleosides.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
69.8 °F - closed cup
Flash Point(C)
21 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Photobromination of (-)-(2R)-2-bromobutane with bromine-81.
Tanner DD, et al.
Journal of the American Chemical Society, 99(8), 2714-2723 (1977)
Selective Reactivity in Gas-Liqiud Chromatography. Determination of 2-Bromobutane and 1-Bromo-2-methylpropane.
Harris WE and McFadden WH.
Analytical Chemistry, 31(1), 114-117 (1959)
Viscosities of binary mixtures of 2-bromobutane and 2-bromo-2-methylpropane with isomeric butanols at 298.15 and 313.15 K.
Artigas H, et al.
International Journal of Thermophysics, 23(6), 1455-1468 (2002)
STEREOCHEMISTRY AND ISOTOPE EFFECT IN THE DEHYDROBROMINATION OF 2-BROMOBUTANE OVER SOLID CATALYSTS.
Misono M and Yoneda Y.
Chemistry Letters (Jpn), 1(7), 551-552 (1972)
Khalid Mohammed Hassan Hilmy
Archiv der Pharmazie, 339(4), 174-181 (2006-04-06)
A series of non-nucleosides 9-47 were synthesized. Compounds 1-4 were reacted with formic acid (85%) to afford compounds 5-8. Then, the latter compounds were reacted with alkyl halides a-f (2-bromopropane, 2-bromobutane, benzyl bromide, benzyl chloromethyl ether, chloromethyl ethyl ether, phenacyl
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service