B78114
2-Bromopropane
99%
Synonym(s):
Isopropyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(CH3)2CHBr
CAS Number:
Molecular Weight:
122.99
Beilstein:
741852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.425 (lit.)
bp
59 °C (lit.)
mp
−89 °C (lit.)
density
1.31 g/mL at 25 °C (lit.)
SMILES string
CC(C)Br
InChI
1S/C3H7Br/c1-3(2)4/h3H,1-2H3
InChI key
NAMYKGVDVNBCFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-Bromopropane is a general reagent used to introduce isopropyl group as in the case of the synthesis of various Buchwald ligands. It can also be used as a starting material in the total synthesis of lamellarin D, lamellarin H, ningalin B and (±)-dichroanal.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Repr. 1A - STOT RE 2 Inhalation
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
66.2 °F - closed cup
Flash Point(C)
19 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The selective reaction of aryl halides with KOH: synthesis of phenols, aromatic ethers, and benzofurans.
Anderson K W, et al.
Journal of the American Chemical Society, 128(33), 10694-10695 (2006)
Efficient route to 4a-methyltetrahydrofluorenes: a total synthesis of (?)-dichroanal B via intramolecular Heck reaction.
Planas L, et al.
The Journal of Organic Chemistry, 71(7), 2896-2898 (2006)
Total synthesis of lamellarins D, H, and R and ningalin B.
Li Q, et al.
Organic Letters, 13(2), 312-315 (2010)
An extremely active catalyst for the Negishi cross-coupling reaction.
Milne J E and Buchwald S L
Journal of the American Chemical Society, 126(40), 13028-13032 (2004)
Katsumi Ohtani et al.
Industrial health, 42(2), 219-225 (2004-05-07)
Reproductive toxicity of 2-bromopropane (2BP), a substitute for ozone layer-depleting chloro-fluorocarbon, was found among the workers in an electronics factory in Korea in 1995. Furthermore the importance of testicular toxicity has been realized since the problem of endocrine disruptors arose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service