Skip to Content
Merck
All Photos(1)

Key Documents

8.20281

Sigma-Aldrich

2-Chlorobutane

for synthesis

Synonym(s):

2-Chlorobutane, sec-Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9Cl
CAS Number:
Molecular Weight:
92.57
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-151-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

160 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

460 °C

potency

17400 mg/kg LD50, oral (Rat)
17400 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-8.8 % (v/v)

pH

7 (20 °C in H2O, Aqueous solution)

bp

65 °C/1013 hPa

mp

-132 °C

transition temp

flash point -10 °C

solubility

1.0 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(CC)C

InChI

1S/C4H9Cl/c1-3-4(2)5/h4H,3H2,1-2H3

InChI key

BSPCSKHALVHRSR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.872 - 0.874
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service