Skip to Content
Merck
All Photos(1)

Key Documents

914657

Sigma-Aldrich

Silica dioxide- vitreous milled nanofiber

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

form

nanofiber

Quality Level

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Inorganic nanofibers have unique properties (i.e. high surface area, high porosity, good breathability, large surface to volume ratio, and stability) their applications include anodes and cathodes in li-ion batteries. The can additionally act as a fuel cell separator, catalyst, catalyst support, photo catalyst, gas sensors, thermal insulators, metal or ceramic nano-composites, dehumidifiers, abrasives, thermal barrier coatings, and filtration.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service