Skip to Content
Merck
All Photos(4)

Key Documents

798568

Sigma-Aldrich

Ammonium formate

anhydrous, free-flowing, Redi-Dri, reagent grade, 97%

Synonym(s):

Formic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
Beilstein:
3625095
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
grade:
anhydrous
reagent grade
form:
crystals
Pricing and availability is not currently available.

grade

anhydrous
reagent grade

Quality Level

product line

Redi-Dri

Assay

97%

form

crystals

mp

119-121 °C (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium formate is the ammonium salt of formic acid. For secondary amines and anilines, ammonium formate is a relatively effective N-formylating agent.

Application

Ammonium formate is used to develop the proton-conducting CMC-AFT (carboxymethyl cellulose - ammonium formate) biopolymer electrolytes system.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service