Skip to Content
Merck
All Photos(1)

Key Documents

5.43834

Supelco

Ammonium acetate

for HPLC LiChropur

Synonym(s):

Ammonium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7NO2
CAS Number:
Molecular Weight:
77.08
MDL number:
UNSPSC Code:
12352106
EC Index Number:
211-162-9
NACRES:
NA.21
Pricing and availability is not currently available.

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

Quality Level

reg. compliance

suitable for FDA C-010.02

form

solid

technique(s)

HPLC: suitable

pH

6.7-7.3 (25 °C, 50 g/L in H2O)

mp

114 °C

solubility

1480 g/L

density

1.17 g/cm3 at 20 °C

bulk density

410 kg/m3

storage temp.

15-25°C

SMILES string

[NH4]OC(=O)C

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

General description

Ammonium acetate is commonly used as a buffer in high-performance liquid chromatography (HPLC). It is used as a mobile phase to develop a novel speciation methodology applicable to urine samples using IEC-ICP-MS.[1]

Application

Ammonium acetate can be used as a:
  • Mobile phase in the derivatization reaction to enhance the detection and separation of glyphosate and aminomethylphosphonic acid (AMPA) using reversed-phase liquid chromatography (RPLC).[2]
  • Buffer component in the mobile phase of the Hydrophilic Interaction Liquid Chromatography (HILIC) method for the separation and determination of polar lipid classes.[3]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium acetate as a novel buffer for highly selective robust urinary HPLC-ICP-MS arsenic speciation methodology
Rodriguez PF, et al.
Talanta, 221, 121494-121494 (2021)
Hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry method for quantification of five phospholipid classes in various matrices
Mazzella N, et al.
MethodsX, 10, 102026-102026 (2023)
Evaluation and optimization of the derivatization reaction conditions of glyphosate and aminomethylphosphonic acid with 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate using reversed-phase liquid chromatography
Fontas, C et al.
Journal of Separation Science, 43(20), 3931-3939 (2020)

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service