Skip to Content
Merck
All Photos(2)

Key Documents

775703

Sigma-Aldrich

Molybdenum(VI) oxide

nanopowder, 100 nm (TEM), 99.5% trace metals basis

Synonym(s):

Molybdenum trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoO3
CAS Number:
Molecular Weight:
143.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5% trace metals basis

form

nanopowder

particle size

100 nm (TEM)

mp

795 °C (lit.)

SMILES string

O=[Mo](=O)=O

InChI

1S/Mo.3O

InChI key

JKQOBWVOAYFWKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Molybdenum oxide nanoparticles have been studied in photocatalytic systems, as gas sensors for automobiles and as anodes in lithium ion batteries.
Precursor to LAMOX fast ion conductors and superconductors.
Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13 and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee, S.H.; et al.
Advanced Materials, 20, 3627-3632 (2008)
Prasad, A. K.; et al.
J. Mater. Sci., 38, 4347-4352 (2003)
Song, K.Y.; et al.
Chemistry of Materials, 13, 2349-2355 (2001)
Yu-Zhan Wang et al.
The Journal of chemical physics, 134(3), 034706-034706 (2011-01-26)
The electronic structures at the MoO(3)∕Co interface were investigated using synchrotron-based ultraviolet and x-ray photoelectron spectroscopy. It was found that interfacial chemical reactions lead to the reduction of Mo oxidation states and the formation of Co-O bonds. These interfacial chemical
Di-Yan Wang et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3415-3420 (2012-06-08)
A heterojunction photodiode with NIR photoresponse using solution processable pyrite FeS(2) nanocrystal ink is demonstrated which has the advantages of earth-abundance and non-toxicity. The device consists of a FeS(2) nanocrystal (NC) thin film sandwiched with semiconducting metal oxides with a

Articles

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service