Skip to Content
Merck
All Photos(1)

Key Documents

774537

Sigma-Aldrich

Tungsten(VI) oxide

nanowires, diam. × L ~50 nm × 10 μm

Synonym(s):

Tungsten trioxide nanowires, Tungstic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WO3
CAS Number:
Molecular Weight:
231.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanowires

diam. × L

~50 nm × 10 μm

density

7.16 g/mL at 25 °C (lit.)

SMILES string

O=[W](=O)=O

InChI

1S/3O.W

InChI key

ZNOKGRXACCSDPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tungsten oxide nanowires have been extensively studied as electrochromics; gas sensors and battery materials.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liao; C.C.;
Solar Energy Mat. and Solar Cells, 90, 1147-1155 (2006)
Polleux; J.;
Angewandte Chemie (International Edition in English), 45, 261-265 (2006)
Gu; Z.;
Journal of Solid State Chemistry, 180, 98-105 (2007)

Articles

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service