Skip to Content
Merck
All Photos(1)

Documents

804169

Sigma-Aldrich

Molybdenum(IV) sulfide

nanopowder, 90 nm diameter (APS), 99% trace metals basis

Synonym(s):

Molybdenum disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MoS2
CAS Number:
Molecular Weight:
160.07
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% trace metals basis

form

nanopowder
powder

diameter

90 nm (APS)

density

5.06 g/mL at 25 °C (lit.)

SMILES string

S=[Mo]=S

InChI

1S/Mo.2S

InChI key

CWQXQMHSOZUFJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MoS2 is an important compound with applications in batteries, lubricants and hydrogen production. The layered structure of MoS2 allows single layers of the compound to be used as 2D nanosheets similar to graphene.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and application of inorganic nanoparticles as lubricant components - A review.
Bakunin VN, et al.
Journal of Nanoparticle Research, 6(2-3), 273-284 (2004)
Jonathan N Coleman et al.
Science (New York, N.Y.), 331(6017), 568-571 (2011-02-05)
If they could be easily exfoliated, layered materials would become a diverse source of two-dimensional crystals whose properties would be useful in applications ranging from electronics to energy storage. We show that layered compounds such as MoS(2), WS(2), MoSe(2), MoTe(2)
Exfoliated MoS2 Nanocomposite as an Anode Material for Lithium Ion Batteries.
Xiao J, et al.
Chemistry of Materials, 22(16), 4522-4524 (2010)
Thomas F Jaramillo et al.
Science (New York, N.Y.), 317(5834), 100-102 (2007-07-07)
The identification of the active sites in heterogeneous catalysis requires a combination of surface sensitive methods and reactivity studies. We determined the active site for hydrogen evolution, a reaction catalyzed by precious metals, on nanoparticulate molybdenum disulfide (MoS2) by atomically

Articles

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Catalytic water splitting produces hydrogen crucial for renewable energy, petroleum refining, and chemical industry applications like methanol production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service