Skip to Content
Merck
All Photos(1)

Documents

76510

Sigma-Aldrich

Pentadecane

≥98.0% (GC)

Synonym(s):

n-Pentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH3
CAS Number:
Molecular Weight:
212.41
Beilstein:
1698194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.4 (vs air)

Quality Level

vapor pressure

1 mmHg ( 91.6 °C)

Assay

≥98.0% (GC)

form

liquid

expl. lim.

6.5 %

refractive index

n20/D 1.431 (lit.)

bp

270 °C (lit.)

mp

8-10 °C (lit.)

density

0.769 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCC

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3

InChI key

YCOZIPAWZNQLMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang-Joon Kim et al.
Journal of hazardous materials, 118(1-3), 171-176 (2005-02-22)
Ex situ electrokinetic (EK) bioremediation of a laboratory-prepared pentadecane-contaminated kaolinite was carried out. Extraneous bacteria and ionic nutrients were continuously supplied to the soil specimen by a new electrolyte circulation method, which controlled electrical pH change of electrolyte solution to
Thembelani E Nomkoko et al.
Dalton transactions (Cambridge, England : 2003), (33)(33), 4029-4038 (2006-10-10)
The equilibrium constants of Cu(II), Zn(II), Ca(II) and Gd(III) with 1,15-bis(N,N-dimethyl)-5,11-dioxo-8-(N-benzyl)-1,4,8,12,15-pentaazapentadecane (La) have been studied at 25 degrees C and an ionic strength of 0.15 mol dm-3. Copper forms more stable complexes with La than the other metal ions investigated.
Heber J Chacon-Madrid et al.
Environmental science & technology, 46(20), 11179-11186 (2012-09-14)
We use a two-dimensional volatility basis set (2D-VBS) box model to simulate secondary organic aerosol (SOA) mass yields of linear oxygenated molecules: n-tridecanal, 2- and 7-tridecanone, 2- and 7-tridecanol, and n-pentadecane. A hybrid model with explicit, a priori treatment of
Kevin M Foote et al.
Organic & biomolecular chemistry, 1(22), 3917-3948 (2003-12-11)
A concise synthesis of the tricyclic furanochroman unit 3 found in the PAF antagonist phomactin A (1) isolated from the marine fungus Phoma sp., is described. In complementary studies, a variety of synthetic routes towards the bicyclo[9.3.1]pentadecane ring system 4
Gina M Geiselman et al.
Microbial cell factories, 19(1), 24-24 (2020-02-07)
Rhodosporidium toruloides has emerged as a promising host for the production of bioproducts from lignocellulose, in part due to its ability to grow on lignocellulosic feedstocks, tolerate growth inhibitors, and co-utilize sugars and lignin-derived monomers. Ent-kaurene derivatives have a diverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service