Skip to Content
Merck
All Photos(1)

Key Documents

733202

Sigma-Aldrich

cis-Dichlorobis(2,2′-bipyridine)ruthenium(II) hydrate

97%

Synonym(s):

cis-Bis(2,2′-bipyridine)dichlororuthenium(II) hydrate, cis-Ru(bpy)2Cl2.xH2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Cl2N4Ru · xH2O
CAS Number:
Molecular Weight:
484.34 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

475.2-493.3

SMILES string

[H]O[H].Cl[Ru]Cl.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4

InChI

1S/2C10H8N2.2ClH.H2O.Ru/c2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;;;;/h2*1-8H;2*1H;1H2;/q;;;;;+2/p-2

InChI key

XBOQABVNRXVAKI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

cis-Dichlorobis(2,2′-bipyridine)ruthenium(II) can be used in synthesizing a variety of ruthenium complexes such as ruthenium(II) complex that can modify the surface of gold nanoparticles for the cysteine detection by selective luminescence. It can also be used in the preparation of ruthenium complexes to design photochromic molecular switches and self-assembling supramolecular systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoinduced electron transfer in a triad that can be assembled/disassembled by two different external inputs. Toward molecular-level electrical extension cables.
Ballardini R, et al.
Journal of the American Chemical Society, 124(43), 12786-12795 (2002)
Simple and selective sensing of cysteine using gold nanoparticles modified by Ruthenium (II) complexes.
Li M J, et al.
Journal of Nanoscience and Nanotechnology, 11(4), 3578-3585 (2011)
Femtosecond isomerization in a photochromic molecular switch.
Mockus N V, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 120(8), 1480-1483 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service