Skip to Content
Merck
All Photos(1)

Key Documents

719536

Sigma-Aldrich

3,3′-Diiodo-L-thyronine (T2) hydrochloride

98% (CP)

Synonym(s):

3,3′-·Diiodo-·4-·(4-·hydroxyphenoxy)·-L-phenylalanine, O-(4-·Hydroxyphenyl)-3-,3′-·diiodo-L-tyrosine, T2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13I2NO4·HCl
CAS Number:
Molecular Weight:
561.54
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

SMILES string

OC1=CC=C(OC2=CC=C(CC(N)C(O)=O)C=C2I)C=C1I.Cl

InChI

1S/C15H13I2NO4/c16-10-7-9(2-3-13(10)19)22-14-4-1-8(5-11(14)17)6-12(18)15(20)21/h1-5,7,12,19H,6,18H2,(H,20,21)/t12-/m0/s1

InChI key

CPCJBZABTUOGNM-LBPRGKRZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debora Bonvin et al.
Dalton transactions (Cambridge, England : 2003), 46(37), 12692-12704 (2017-09-16)
The overexpression of the folate receptor in most cancers has been widely exploited to specifically deliver folic acid (FA) coupled nanomedicines to tumors. However, complex coupling chemistry is often used to bind FA to the nanoparticles. Furthermore, very little has
Viviane G Nasr et al.
Anesthesia and analgesia, 128(4), 661-668 (2018-01-13)
Cerebral oximetry using near-infrared spectroscopy is a noninvasive optical technology to detect cerebral hypoxia-ischemia and develop interventions to prevent and ameliorate hypoxic brain injury. Cerebral oximeters are calibrated and validated by comparison of the near-infrared spectroscopy-measured cerebral O2 saturation (SctO2)
Satoshi Tsutsumi et al.
Surgical and radiologic anatomy : SRA, 41(9), 1045-1051 (2019-07-18)
There has not been a study documenting the distribution of cerebrospinal fluid (CSF) pathways in the anterolateral base of the middle fossa (ALB) and diploe of the pterional region (Pt). The present study aimed to delineate these pathways using magnetic
Ahmed Abdel Khalek Abdel Razek et al.
Journal of computer assisted tomography, 43(3), 467-471 (2019-05-15)
This study aimed to assess metrics of diffusion tensor imaging in evaluating microstructural abnormalities of the lateral rectus muscle in Duane retraction syndrome (DRS). A prospective study was conducted on 27 patients with DRS and 16 age- and sex-matched controls
Kenneth H Mayer et al.
PLoS medicine, 14(11), e1002435-e1002435 (2017-11-15)
VRC01 is an HIV-1 CD4 binding site broadly neutralizing antibody (bnAb) that is active against a broad range of HIV-1 primary isolates in vitro and protects against simian-human immunodeficiency virus (SHIV) when delivered parenterally to nonhuman primates. It has been

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service