54680
Butyl 4-hydroxybenzoate
≥99.0% (GC)
Synonym(s):
p-Hydroxybenzoic acid n-butyl ester, Butyl paraben
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
194.23
Beilstein:
1103741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99.0% (GC)
bp
156-157 °C/3.5 mmHg (lit.)
mp
67-70 °C (lit.)
68-70 °C
functional group
ester
SMILES string
CCCCOC(=O)c1ccc(O)cc1
InChI
1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3
InChI key
QFOHBWFCKVYLES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Butyl 4-hydroxybenzoate can be used to synthesize 4-[(9-oxoxanthen-3-yl)-carbonyloxy]benzoate by reacting with 2-phenoxyterephthaloyl dichloride in hot pyridine.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
3-(4-Butoxycarbonylphenyloxycarbonyl)-xanthen-9-one.
Centore, R, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 51(6), 1211-1213 (1995)
Guoyuan Wen et al.
The Journal of general virology, 96(Pt 6), 1219-1228 (2015-01-30)
Thermostable Newcastle disease virus (NDV) vaccines have been used widely to control Newcastle disease for village poultry flocks, due to their independence of cold chains for delivery and storage. To explore the potential use of thermostable NDV as a vaccine
Jordan Shaw et al.
Reproductive toxicology (Elmsford, N.Y.), 28(1), 26-31 (2009-06-06)
Parabens, a class of preservatives routinely added to cosmetics, pharmaceuticals, and foods, have estrogenic properties. Given that intrauterine implantation of fertilized ova in inseminated females can be disrupted by minute levels of exogenous estrogens, we assessed the impact of parabens
Caroline Stremnitzer et al.
The Journal of investigative dermatology, 135(7), 1790-1800 (2015-02-24)
Papain is commonly used in food, pharmaceutical, textile, and cosmetic industries and is known to induce occupational allergic asthma. We have previously shown that the papain-like cysteine protease Dermatophagoides pteronyssinus 1 from house dust mite exhibits percutaneous sensitization potential. We
Emeran A Mayer et al.
The Journal of clinical investigation, 125(3), 926-938 (2015-02-18)
Tremendous progress has been made in characterizing the bidirectional interactions between the central nervous system, the enteric nervous system, and the gastrointestinal tract. A series of provocative preclinical studies have suggested a prominent role for the gut microbiota in these
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service