Skip to Content
Merck
All Photos(3)

Documents

682691

Sigma-Aldrich

Bis(tri-tert-butylphosphine)platinum(0)

Synonym(s):

Bis[tris(1,1-dimethylethyl)phosphine]platinum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[((CH3)3C)3P]2Pt
CAS Number:
Molecular Weight:
599.71
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

mp

260-269 °C

storage temp.

2-8°C

SMILES string

[Pt].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/2C12H27P.Pt/c2*1-10(2,3)13(11(4,5)6)12(7,8)9;/h2*1-9H3;

InChI key

RJQWVEJVXWLMRE-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Trimetallic ReBiPt complexes
  • Rhodium-germanium carbonyl complexes

Catalyst for the synthesis of:
  • Amidobipyridyl inhibitors of protein kinase D
  • 5-Fluoroalkylated pyrimidine nucleosides via Negishi cross-coupling

Catalyst for phenyl and methyl group transfer from tin to iridium

Activator of metal hydride complexes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service