Skip to Content
Merck
All Photos(4)

Key Documents

479519

Sigma-Aldrich

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution

in xylene, Pt ~2 %

Synonym(s):

Karstedt′s catalyst, Platinum(0) 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O[Si(CH3)2CH=CH2]2Pt
CAS Number:
Molecular Weight:
381.48
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7 mmHg ( 21 °C)

composition

Pt, ~2%

reaction suitability

core: platinum
reagent type: catalyst

concentration

in xylene

bp

138 °C

mp

12-13 °C

density

0.855 g/mL at 25 °C

SMILES string

[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C

InChI

1S/C8H18OSi2.Pt/c1-7-10(3,4)9-11(5,6)8-2;/h7-8H,1-2H2,3-6H3;

InChI key

RCNRJBWHLARWRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex is a platinum complex, commonly known as a Karstedt′s catalyst. It is generally employed as a catalyst in hydrosilylation reactions.

Application

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution is a powerful hydrosilylation catalyst for the preparation of trans-β-silanes.

It has also been used in the reduction of carboxamides to amines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platinum (0)-1, 3-divinyl-1, 1, 3, 3-tetramethyldisiloxane Complex as a Pt Source for Pt/SnO2 Catalyst
Martyla A, et al.
Journal of Nanomaterials, 2014 (2014)
Practical access to amines by platinum-catalyzed reduction of carboxamides with hydrosilanes: synergy of dual Si- H groups leads to high efficiency and selectivity
Hanada S, et al.
Journal of the American Chemical Society, 131, 15032-15040 (2009)
Kenichiro Itami et al.
The Journal of organic chemistry, 67(8), 2645-2652 (2002-04-13)
Metal-catalyzed hydrosilylation of alkenes and alkynes using dimethyl(pyridyl)silane is described. The hydrosilylation of alkenes using dimethyl(2-pyridyl)silane (2-PyMe(2)SiH) proceeded well in the presence of a catalytic amount of RhCl(PPh(3))(3) with virtually complete regioselectivity. By taking advantage of the phase tag property
Denmark, S. E.; Wang, Z.
Organic Syntheses, 81, 54-54 (2005)
S E Denmark et al.
Organic letters, 3(7), 1073-1076 (2001-03-30)
[reaction: see text]. The formal addition of an aryl-H or alkenyl-H bond across a terminal alkyne has been accomplished by the combination of platinum-catalyzed hydrosilylation followed by palladium-catalyzed cross-coupling. The use of the t-Bu3P-Pt(DVDS) catalyst in combination with tetramethyldisiloxane gave

Articles

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service